Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCTGGGCAGGAGCCTCTGGTTC[C/T]CCCGGGGTGGGAGGCCCCAGCCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606383 MIM: 609124 | ||||||||||||||||||||
Literature Links: |
GPR84 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR84 - G protein-coupled receptor 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC102724050 - uncharacterized LOC102724050 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF385A - zinc finger protein 385A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130967.2 | 971 | Silent Mutation | NP_001124439.1 | |||
NM_001130968.2 | 971 | Silent Mutation | NP_001124440.1 | |||
NM_001290001.1 | 971 | Silent Mutation | NP_001276930.1 | |||
NM_001290002.1 | 971 | Silent Mutation | NP_001276931.1 | |||
NM_001290004.1 | 971 | Silent Mutation | NP_001276933.1 | |||
NM_015481.2 | 971 | Silent Mutation | NP_056296.1 | |||
XM_005268783.4 | 971 | Silent Mutation | XP_005268840.1 | |||
XM_006719342.2 | 971 | Silent Mutation | XP_006719405.1 | |||
XM_011538168.2 | 971 | Silent Mutation | XP_011536470.1 | |||
XM_011538169.2 | 971 | Silent Mutation | XP_011536471.1 | |||
XM_011538170.1 | 971 | Silent Mutation | XP_011536472.1 | |||
XM_011538171.2 | 971 | Silent Mutation | XP_011536473.1 | |||
XM_017019174.1 | 971 | Silent Mutation | XP_016874663.1 |