Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGCTGTGGGGTAGGTTCGGGT[A/G]CCCGGTTTGCCAGGCGGCTGAGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603277 MIM: 164031 | ||||||||||||||||||||
Literature Links: |
CHD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHD4 - chromodomain helicase DNA binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001273.3 | 5803 | Missense Mutation | GCA,GTA | A,V 1900 | NP_001264.2 | |
NM_001297553.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1893 | NP_001284482.1 | |
XM_005253668.3 | 5803 | Missense Mutation | GCA,GTA | A,V 1893 | XP_005253725.1 | |
XM_006718958.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1928 | XP_006719021.1 | |
XM_006718959.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1903 | XP_006719022.1 | |
XM_006718960.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1902 | XP_006719023.1 | |
XM_006718961.2 | 5803 | Missense Mutation | GCA,GTA | A,V 1896 | XP_006719024.1 | |
XM_006718962.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1890 | XP_006719025.1 | |
XM_017018725.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1902 | XP_016874214.1 | |
XM_017018726.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1901 | XP_016874215.1 | |
XM_017018727.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1900 | XP_016874216.1 | |
XM_017018728.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1899 | XP_016874217.1 | |
XM_017018729.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1894 | XP_016874218.1 | |
XM_017018730.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1888 | XP_016874219.1 | |
XM_017018731.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1887 | XP_016874220.1 | |
XM_017018732.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1883 | XP_016874221.1 | |
XM_017018733.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1881 | XP_016874222.1 | |
XM_017018734.1 | 5803 | Missense Mutation | GCA,GTA | A,V 1880 | XP_016874223.1 |
NOP2 - NOP2 nucleolar protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |