Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTAGGTTTTAGGCTGAAGA[C/T]GTAAAGCCTGCTGCTCCTGGGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606634 MIM: 607360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCD - dermcidin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300854.1 | 571 | UTR 3 | NP_001287783.1 | |||
NM_053283.3 | 571 | UTR 3 | NP_444513.1 |
LACRT - lacritin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |