Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCATGGAGCCCTCCTTCACCA[C/T]GCGCTCCTTGAGCACGGGGGCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616569 | ||||||||||||||||||||
Literature Links: |
CSAD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSAD - cysteine sulfinic acid decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244705.1 | 1218 | Missense Mutation | ATG,GTG | M,V 445 | NP_001231634.1 | |
NM_001244706.1 | 1218 | Intron | NP_001231635.1 | |||
NM_015989.4 | 1218 | Missense Mutation | ATG,GTG | M,V 472 | NP_057073.4 | |
XM_011538442.2 | 1218 | Missense Mutation | ATG,GTG | M,V 503 | XP_011536744.2 | |
XM_011538446.2 | 1218 | Missense Mutation | ATG,GTG | M,V 472 | XP_011536748.1 | |
XM_011538449.1 | 1218 | Missense Mutation | ATG,GTG | M,V 245 | XP_011536751.1 | |
XM_011538451.1 | 1218 | Missense Mutation | ATG,GTG | M,V 227 | XP_011536753.1 | |
XM_017019416.1 | 1218 | Intron | XP_016874905.1 | |||
XM_017019417.1 | 1218 | Missense Mutation | ATG,GTG | M,V 245 | XP_016874906.1 | |
XM_017019418.1 | 1218 | Missense Mutation | ATG,GTG | M,V 227 | XP_016874907.1 | |
XM_017019419.1 | 1218 | Missense Mutation | ATG,GTG | M,V 320 | XP_016874908.1 |