Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACACGGCTGAAGCCACCCCACAA[A/G]CTGCACCCAAACTTCACTTCACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602692 | ||||||||||||||||||||
Literature Links: |
GLIPR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLIPR1 - GLI pathogenesis related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006851.2 | 391 | Silent Mutation | AAA,AAG | K,K 88 | NP_006842.2 | |
XM_011537775.2 | 391 | Silent Mutation | AAA,AAG | K,K 88 | XP_011536077.1 | |
XM_017018721.1 | 391 | UTR 5 | XP_016874210.1 |