Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCTGGGACATCCCCTGCCTCAA[C/T]GCACGATAATTTTGCCTTTGGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHG6 - pleckstrin homology and RhoGEF domain containing G6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144856.1 | 311 | Intron | NP_001138328.1 | |||
NM_001144857.1 | 311 | UTR 5 | NP_001138329.1 | |||
NM_018173.3 | 311 | Intron | NP_060643.2 | |||
XM_005253704.4 | 311 | Intron | XP_005253761.1 | |||
XM_006718985.3 | 311 | Intron | XP_006719048.1 | |||
XM_011520967.2 | 311 | Intron | XP_011519269.1 | |||
XM_017019555.1 | 311 | Intron | XP_016875044.1 |