Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCACTGACCCACACTTTATAG[A/G]CAACTGCATAGAAGCCCACAACGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607724 MIM: 610395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIPR1L1 - GLI pathogenesis related 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304964.1 | 2999 | Missense Mutation | GAC,GGC | D,G 35 | NP_001291893.1 | |
NM_152779.3 | 2999 | Missense Mutation | GAC,GGC | D,G 35 | NP_689992.1 | |
XM_011538134.2 | 2999 | Missense Mutation | GAC,GGC | D,G 120 | XP_011536436.2 | |
XM_017019134.1 | 2999 | Missense Mutation | GAC,GGC | D,G 120 | XP_016874623.1 |