Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAAATCAGCTGAAAGACATCCA[A/C]AGTCATGGTATCATTCAGAATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608722 MIM: 608075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPZA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPZA3 - capping actin protein of muscle Z-line alpha subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033328.2 | 473 | Missense Mutation | CAA,CAC | Q,H 105 | NP_201585.1 |
PLCZ1 - phospholipase C zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033123.3 | 473 | Intron | NP_149114.2 | |||
XM_005253519.2 | 473 | Intron | XP_005253576.1 | |||
XM_011520883.1 | 473 | Intron | XP_011519185.1 | |||
XM_017020176.1 | 473 | Intron | XP_016875665.1 | |||
XM_017020177.1 | 473 | Intron | XP_016875666.1 | |||
XM_017020178.1 | 473 | Intron | XP_016875667.1 | |||
XM_017020179.1 | 473 | Intron | XP_016875668.1 | |||
XM_017020180.1 | 473 | Intron | XP_016875669.1 | |||
XM_017020181.1 | 473 | Intron | XP_016875670.1 | |||
XM_017020182.1 | 473 | Intron | XP_016875671.1 | |||
XM_017020183.1 | 473 | Intron | XP_016875672.1 | |||
XM_017020184.1 | 473 | Intron | XP_016875673.1 | |||
XM_017020185.1 | 473 | Intron | XP_016875674.1 | |||
XM_017020186.1 | 473 | Intron | XP_016875675.1 |