Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGTCGAGCGGCCCGTGTCGCG[C/T]ATGGGCCAAAAGCAGGAGAACGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605476 MIM: 609677 | ||||||||||||||||||||
Literature Links: |
AGAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGAP2 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122772.2 | 2351 | Missense Mutation | ACG,GCG | T,A 1077 | NP_001116244.1 | |
NM_014770.3 | 2351 | Missense Mutation | ACG,GCG | T,A 721 | NP_055585.1 | |
XM_005268625.3 | 2351 | Missense Mutation | ACG,GCG | T,A 1057 | XP_005268682.1 | |
XM_005268626.2 | 2351 | Missense Mutation | ACG,GCG | T,A 741 | XP_005268683.1 | |
XM_017018772.1 | 2351 | Missense Mutation | ACG,GCG | T,A 1077 | XP_016874261.1 |
AGAP2-AS1 - AGAP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OS9 - OS9, endoplasmic reticulum lectin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |