Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACTACCACCATGTCACCAAGCT[C/G]GACGAGGTGGCTGCCTCTCAGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASIC1 - acid sensing ion channel subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001095.3 | 310 | Silent Mutation | CTC,CTG | L,L 77 | NP_001086.2 | |
NM_001256830.1 | 310 | Intron | NP_001243759.1 | |||
NM_020039.3 | 310 | Silent Mutation | CTC,CTG | L,L 77 | NP_064423.2 | |
XM_011538350.1 | 310 | Silent Mutation | CTC,CTG | L,L 77 | XP_011536652.1 | |
XM_011538351.2 | 310 | Silent Mutation | CTC,CTG | L,L 77 | XP_011536653.1 | |
XM_011538352.1 | 310 | Silent Mutation | CTC,CTG | L,L 77 | XP_011536654.1 |