Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTCCAGCATTATATTGAAACACT[A/G]CAGCGCAGGATTGAAGAAGAGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176876 | ||||||||||||||||||||
Literature Links: |
PTPN11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPN11 - protein tyrosine phosphatase, non-receptor type 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002834.3 | 1955 | Silent Mutation | NP_002825.3 | |||
XM_006719526.2 | 1955 | Intron | XP_006719589.1 | |||
XM_011538613.2 | 1955 | Intron | XP_011536915.1 | |||
XM_017019722.1 | 1955 | Intron | XP_016875211.1 |