Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCCAGGGCCAATTTGAGGTTTC[A/C]GTGTTTGTGCCTGAACCATCGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMKLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMKLR1 - chemerin chemokine-like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142343.1 | 4529 | UTR 3 | NP_001135815.1 | |||
NM_001142344.1 | 4529 | UTR 3 | NP_001135816.1 | |||
NM_001142345.1 | 4529 | UTR 3 | NP_001135817.1 | |||
NM_004072.2 | 4529 | UTR 3 | NP_004063.1 | |||
XM_017018820.1 | 4529 | UTR 3 | XP_016874309.1 |