Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTAGCTCACTGTTGTCAAAAGAA[A/G]TGGTATAGGACAATGGTGGTAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAP1L1 - nucleosome assembly protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719425.3 | Intron | XP_006719488.1 | ||||
XM_011538390.2 | Intron | XP_011536692.2 | ||||
XM_011538393.2 | Intron | XP_011536695.1 | ||||
XM_017019338.1 | Intron | XP_016874827.1 | ||||
XM_017019339.1 | Intron | XP_016874828.1 | ||||
XM_017019340.1 | Intron | XP_016874829.1 |