Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAAGGGCGCCCTTCCCCTGGAT[A/G]CGGTCACTTTCTACAAGGTAACCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602287 | ||||||||||||||||||||
Literature Links: |
ERP29 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERP29 - endoplasmic reticulum protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034025.1 | 245 | Missense Mutation | ACG,GCG | T,A 43 | NP_001029197.1 | |
NM_006817.3 | 245 | Missense Mutation | ACG,GCG | T,A 43 | NP_006808.1 | |
XM_017018720.1 | 245 | Intron | XP_016874209.1 |