Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAAAGCTTCACCTTTCTCCAAA[C/G]GCAGATGTGAAGAACTTGATGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LETMD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LETMD1 - LETM1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243689.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | NP_001230618.1 | |
NM_001300765.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | NP_001287694.1 | |
NM_015416.4 | 208 | Missense Mutation | AAC,AAG | N,K 50 | NP_056231.3 | |
XM_006719335.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_006719398.1 | |
XM_006719336.1 | 208 | UTR 5 | XP_006719399.1 | |||
XM_006719339.1 | 208 | UTR 5 | XP_006719402.1 | |||
XM_006719340.1 | 208 | UTR 5 | XP_006719403.1 | |||
XM_011538161.2 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_011536463.1 | |
XM_011538162.2 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_011536464.1 | |
XM_011538163.2 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_011536465.1 | |
XM_011538164.2 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_011536466.1 | |
XM_011538165.2 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_011536467.1 | |
XM_011538167.1 | 208 | Intron | XP_011536469.1 | |||
XM_017019151.1 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_016874640.1 | |
XM_017019152.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874641.1 | |
XM_017019153.1 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_016874642.1 | |
XM_017019154.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874643.1 | |
XM_017019155.1 | 208 | UTR 5 | XP_016874644.1 | |||
XM_017019156.1 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_016874645.1 | |
XM_017019157.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874646.1 | |
XM_017019158.1 | 208 | UTR 5 | XP_016874647.1 | |||
XM_017019159.1 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_016874648.1 | |
XM_017019160.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874649.1 | |
XM_017019161.1 | 208 | Missense Mutation | AAC,AAG | N,K 57 | XP_016874650.1 | |
XM_017019162.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874651.1 | |
XM_017019163.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874652.1 | |
XM_017019164.1 | 208 | UTR 5 | XP_016874653.1 | |||
XM_017019165.1 | 208 | UTR 5 | XP_016874654.1 | |||
XM_017019166.1 | 208 | UTR 5 | XP_016874655.1 | |||
XM_017019167.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874656.1 | |
XM_017019168.1 | 208 | Missense Mutation | AAC,AAG | N,K 50 | XP_016874657.1 |