Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCAGCCTCTGTGCATTGGTTCC[A/G]GAACCGGGGCCAGGGCCGAGTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153337 MIM: 168440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAG3 - lymphocyte activating 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002286.5 | 981 | Missense Mutation | CAG,CGG | Q,R 204 | NP_002277.4 | |
XM_011520956.1 | 981 | Missense Mutation | CAG,CGG | Q,R 204 | XP_011519258.1 |
PTMS - parathymosin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |