Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGTGGATGGGCAGGAGGGTGGC[C/T]GGGTTAACAGAGGGTGTGGTAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP54 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP54 - cilia and flagella associated protein 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306084.1 | 225 | Intron | NP_001293013.1 | |||
XM_011539069.2 | 225 | Intron | XP_011537371.1 | |||
XM_011539070.2 | 225 | Intron | XP_011537372.1 | |||
XM_011539071.2 | 225 | Intron | XP_011537373.1 | |||
XM_011539072.2 | 225 | Intron | XP_011537374.1 | |||
XM_011539073.2 | 225 | Intron | XP_011537375.1 | |||
XM_011539074.2 | 225 | Intron | XP_011537376.1 | |||
XM_011539076.2 | 225 | Intron | XP_011537378.1 | |||
XM_011539077.2 | 225 | UTR 5 | XP_011537379.1 | |||
XM_011539078.2 | 225 | Intron | XP_011537380.1 | |||
XM_011539079.1 | 225 | Intron | XP_011537381.1 | |||
XM_011539081.2 | 225 | Intron | XP_011537383.1 | |||
XM_011539083.2 | 225 | Intron | XP_011537385.1 | |||
XM_011539084.2 | 225 | Intron | XP_011537386.1 | |||
XM_011539085.1 | 225 | Intron | XP_011537387.1 | |||
XM_017018866.1 | 225 | Intron | XP_016874355.1 | |||
XM_017018867.1 | 225 | Intron | XP_016874356.1 |