Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACCCAGCCCCCTCCCTCTCCT[C/G]CCCACATCCAAGAACAACAGAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116953 MIM: 155550 MIM: 179514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK2 - cyclin dependent kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290230.1 | Intron | NP_001277159.1 | ||||
NM_001798.4 | Intron | NP_001789.2 | ||||
NM_052827.3 | Intron | NP_439892.2 | ||||
XM_011537732.1 | Intron | XP_011536034.1 |
PMEL - premelanosome protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |