Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGCCACTCCTGCTATTCAACTG[C/T]CATCTTCTTTATATGATGAAGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
UHRF1BP1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UHRF1BP1L - UHRF1 binding protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006947.1 | 3088 | Intron | NP_001006948.1 | |||
NM_015054.1 | 3088 | Missense Mutation | ACA,GCA | T,A 1462 | NP_055869.1 | |
XM_005268738.2 | 3088 | Missense Mutation | ACA,GCA | T,A 1414 | XP_005268795.1 | |
XM_005268739.3 | 3088 | Missense Mutation | ACA,GCA | T,A 1401 | XP_005268796.1 | |
XM_011538054.2 | 3088 | Missense Mutation | ACA,GCA | T,A 980 | XP_011536356.1 | |
XM_017019047.1 | 3088 | Intron | XP_016874536.1 | |||
XM_017019048.1 | 3088 | Missense Mutation | ACA,GCA | T,A 1353 | XP_016874537.1 | |
XM_017019049.1 | 3088 | Intron | XP_016874538.1 | |||
XM_017019050.1 | 3088 | Missense Mutation | ACA,GCA | T,A 1009 | XP_016874539.1 | |
XM_017019051.1 | 3088 | Missense Mutation | ACA,GCA | T,A 1009 | XP_016874540.1 |