Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCAGCAACGCCAAGAAGCAGGC[A/T]CTGCATGCCGTCATGGGCATCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186940 MIM: 610342 | ||||||||||||||||||||
Literature Links: |
CD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD4 - CD4 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR162 - G protein-coupled receptor 162 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014449.1 | 937 | Intron | NP_055264.1 | |||
NM_019858.1 | 937 | Silent Mutation | GCA,GCT | A,A 134 | NP_062832.1 |
P3H3 - prolyl 3-hydroxylase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |