Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCCTTCTTTTGTGGGTCCTTAG[A/G]TCCTGGGAAGGAAATGAGTGACAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 134921 | ||||||||||||||||||||
Literature Links: |
FGF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGF6 - fibroblast growth factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020996.2 | 667 | Missense Mutation | ACC,ATC | T,I 208 | NP_066276.2 | |
XM_017018995.1 | 667 | Intron | XP_016874484.1 | |||
XM_017018996.1 | 667 | Intron | XP_016874485.1 |