Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTACCTGGTCTCCAGAATGGAC[A/G]GCCCTGTGGCAGAGCATGCCAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SDSL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDSL - serine dehydratase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304993.1 | 651 | Missense Mutation | AGC,GGC | S,G 3 | NP_001291922.1 | |
NM_138432.3 | 651 | Missense Mutation | AGC,GGC | S,G 3 | NP_612441.1 | |
XM_005253831.4 | 651 | Missense Mutation | AGC,GGC | S,G 3 | XP_005253888.1 | |
XM_011537846.2 | 651 | Missense Mutation | AGC,GGC | S,G 3 | XP_011536148.1 | |
XM_017018763.1 | 651 | Missense Mutation | AGC,GGC | S,G 16 | XP_016874252.1 |