Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGGTAGTCGAAGCTGGCACGC[A/G]ACAGGGCCTTGACCCAGCCCTCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614239 | ||||||||||||||||||||
Literature Links: |
FAM109A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM109A - family with sequence similarity 109 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177996.1 | 717 | Missense Mutation | TCG,TTG | S,L 122 | NP_001171467.1 | |
NM_001177997.1 | 717 | Missense Mutation | TCG,TTG | S,L 109 | NP_001171468.1 | |
NM_144671.4 | 717 | Missense Mutation | TCG,TTG | S,L 109 | NP_653272.2 | |
XM_006719257.3 | 717 | Missense Mutation | TCG,TTG | S,L 109 | XP_006719320.1 | |
XM_011537976.2 | 717 | Missense Mutation | TCG,TTG | S,L 109 | XP_011536278.1 | |
XM_017018883.1 | 717 | Missense Mutation | TCG,TTG | S,L 109 | XP_016874372.1 |