Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCGGGGAGCCTGGCTTGGGGAC[A/G]TCTATATCTGAGGGAGAAGAGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602341 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXM1 - forkhead box M1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243088.1 | 2377 | Silent Mutation | GAC,GAT | D,D 683 | NP_001230017.1 | |
NM_001243089.1 | 2377 | Silent Mutation | GAC,GAT | D,D 682 | NP_001230018.1 | |
NM_021953.3 | 2377 | Silent Mutation | GAC,GAT | D,D 698 | NP_068772.2 | |
NM_202002.2 | 2377 | Silent Mutation | GAC,GAT | D,D 736 | NP_973731.1 | |
NM_202003.2 | 2377 | Silent Mutation | GAC,GAT | D,D 683 | NP_973732.1 | |
XM_005253676.3 | 2377 | Silent Mutation | GAC,GAT | D,D 699 | XP_005253733.1 | |
XM_011520930.2 | 2377 | Silent Mutation | GAC,GAT | D,D 698 | XP_011519232.1 | |
XM_011520931.2 | 2377 | Silent Mutation | GAC,GAT | D,D 684 | XP_011519233.1 | |
XM_011520932.1 | 2377 | Intron | XP_011519234.1 | |||
XM_011520933.1 | 2377 | Intron | XP_011519235.1 | |||
XM_011520934.2 | 2377 | UTR 3 | XP_011519236.1 | |||
XM_011520935.1 | 2377 | Intron | XP_011519237.1 |
LOC100507424 - uncharacterized LOC100507424 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929469 - uncharacterized protein ENSP00000372125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |