Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGAAGGGATTTCTGAAATGCCA[A/G]CGTAGTTTTTTTTGGCCATCCTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602927 | ||||||||||||||||||||
Literature Links: |
GPR19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR19 - G protein-coupled receptor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006143.2 | 3813 | Missense Mutation | GCT,GTT | A,V 372 | NP_006134.1 | |
XM_011520623.2 | 3813 | Missense Mutation | GCT,GTT | A,V 391 | XP_011518925.1 | |
XM_011520624.2 | 3813 | Missense Mutation | GCT,GTT | A,V 391 | XP_011518926.1 | |
XM_011520625.2 | 3813 | Missense Mutation | GCT,GTT | A,V 372 | XP_011518927.1 | |
XM_017019212.1 | 3813 | Missense Mutation | GCT,GTT | A,V 391 | XP_016874701.1 | |
XM_017019213.1 | 3813 | Missense Mutation | GCT,GTT | A,V 391 | XP_016874702.1 | |
XM_017019214.1 | 3813 | Missense Mutation | GCT,GTT | A,V 391 | XP_016874703.1 | |
XM_017019215.1 | 3813 | Missense Mutation | GCT,GTT | A,V 391 | XP_016874704.1 | |
XM_017019216.1 | 3813 | Missense Mutation | GCT,GTT | A,V 372 | XP_016874705.1 |