Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCATTCCAAAGGGCCATGTAAA[C/G]TTCTTCGCGATTGCACGGATTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC77 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC77 - coiled-coil domain containing 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130146.1 | 4 | UTR 5 | NP_001123618.1 | |||
NM_001130147.1 | 4 | Intron | NP_001123619.1 | |||
NM_001130148.1 | 4 | Intron | NP_001123620.1 | |||
NM_032358.3 | 4 | UTR 5 | NP_115734.1 |