Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCTTCTGCTGGCCTCTCTGCTC[G/T]CGTCCTGCTCCTGTAACAAAGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611324 MIM: 611866 | ||||||||||||||||||||
Literature Links: |
C1RL-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1RL-AS1 - C1RL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLSTN3 - calsyntenin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014718.3 | 321 | Missense Mutation | GCG,TCG | A,S 15 | NP_055533.2 | |
XM_006719163.3 | 321 | Intron | XP_006719226.1 |
RBP5 - retinol binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017020002.1 | 321 | Intron | XP_016875491.1 |