Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTGTCCCTCCTGGATTTCCTGC[C/T]GGGTCTCGGCAGGCAGATGGTTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604986 | ||||||||||||||||||||
Literature Links: |
BRAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRAP - BRCA1 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006768.4 | 1656 | Missense Mutation | NP_006759.3 | |||
XM_005253944.4 | 1656 | Missense Mutation | XP_005254001.1 | |||
XM_011538789.2 | 1656 | Missense Mutation | XP_011537091.1 | |||
XM_017019992.1 | 1656 | Missense Mutation | XP_016875481.1 | |||
XM_017019993.1 | 1656 | Missense Mutation | XP_016875482.1 | |||
XM_017019994.1 | 1656 | Missense Mutation | XP_016875483.1 | |||
XM_017019995.1 | 1656 | Missense Mutation | XP_016875484.1 |