Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTTTCACAGACATAATCCCGT[C/T]TTCCTTCATGAGAATTGAGATGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605780 | ||||||||||||||||||||
Literature Links: |
LOC101929162 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929162 - uncharacterized LOC101929162 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDM4 - PR domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012406.3 | 2617 | Missense Mutation | AAA,AGA | K,R 727 | NP_036538.3 | |
XM_005268593.1 | 2617 | Missense Mutation | AAA,AGA | K,R 734 | XP_005268650.1 |