Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGGCGGGTGTTGTGAGCGGGA[A/G]GGTAGGTAACGGCCTCCAGGAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRS2P2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRS2P2 - MRS2 pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D15 - TBC1 domain family member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146213.1 | 104 | Missense Mutation | AAG,AGG | K,R 10 | NP_001139685.2 | |
NM_001146214.1 | 104 | UTR 5 | NP_001139686.1 | |||
NM_022771.4 | 104 | Missense Mutation | AAG,AGG | K,R 10 | NP_073608.4 | |
XM_006719564.2 | 104 | Intron | XP_006719627.1 | |||
XM_006719565.3 | 104 | Intron | XP_006719628.1 | |||
XM_006719566.2 | 104 | UTR 5 | XP_006719629.1 | |||
XM_006719567.2 | 104 | UTR 5 | XP_006719630.1 | |||
XM_011538679.1 | 104 | UTR 5 | XP_011536981.1 | |||
XM_011538680.1 | 104 | Intron | XP_011536982.1 | |||
XM_011538681.2 | 104 | Missense Mutation | AAG,AGG | K,R 10 | XP_011536983.1 | |
XM_011538682.1 | 104 | UTR 5 | XP_011536984.1 | |||
XM_017019824.1 | 104 | UTR 5 | XP_016875313.1 | |||
XM_017019825.1 | 104 | UTR 5 | XP_016875314.1 | |||
XM_017019826.1 | 104 | UTR 5 | XP_016875315.1 | |||
XM_017019827.1 | 104 | Intron | XP_016875316.1 | |||
XM_017019828.1 | 104 | UTR 5 | XP_016875317.1 |