Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCAGTCCGTTTGTCTAGATGAC[A/G]CAACAGTGAAGTTTGAGATCTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179514 MIM: 606887 | ||||||||||||||||||||
Literature Links: |
RAB5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252036.1 | 420 | Missense Mutation | ACA,GCA | T,A 67 | NP_001238965.1 | |
NM_001252037.1 | 420 | Missense Mutation | ACA,GCA | T,A 67 | NP_001238966.1 | |
NM_002868.3 | 420 | Missense Mutation | ACA,GCA | T,A 67 | NP_002859.1 | |
XM_005269051.2 | 420 | Missense Mutation | ACA,GCA | T,A 67 | XP_005269108.1 | |
XM_005269052.2 | 420 | Missense Mutation | ACA,GCA | T,A 67 | XP_005269109.1 |
SUOX - sulfite oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |