Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTCCGCGGAGGCGGCTCCAAGG[A/G]AGCATTTGGGGTCCCAGCTGATTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123805 | ||||||||||||||||||||
Literature Links: |
PDE3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE3A - phosphodiesterase 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000921.4 | 845 | Missense Mutation | AAG,GAG | K,E 269 | NP_000912.3 | |
NM_001244683.1 | 845 | Intron | NP_001231612.1 | |||
XM_017019420.1 | 845 | Missense Mutation | AAG,GAG | K,E 269 | XP_016874909.1 | |
XM_017019421.1 | 845 | Intron | XP_016874910.1 |