Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGGTGCAATGTCAATGATGGG[C/G]AGGCGTGAGGTCTTCTGTGACCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120140 | ||||||||||||||||||||
Literature Links: |
COL2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL2A1 - collagen type II alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001844.4 | 5946 | Silent Mutation | CTC,CTG | L,L 1460 | NP_001835.3 | |
NM_033150.2 | 5946 | Silent Mutation | CTC,CTG | L,L 1391 | NP_149162.2 | |
XM_017018828.1 | 5946 | Silent Mutation | CTC,CTG | L,L 1508 | XP_016874317.1 | |
XM_017018829.1 | 5946 | Silent Mutation | CTC,CTG | L,L 1507 | XP_016874318.1 | |
XM_017018830.1 | 5946 | Silent Mutation | CTC,CTG | L,L 1438 | XP_016874319.1 | |
XM_017018831.1 | 5946 | Silent Mutation | CTC,CTG | L,L 1278 | XP_016874320.1 |
TMEM106C - transmembrane protein 106C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |