Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTACCTGAAACATTTGCATCCCT[A/G]TTATATCTAGGGAACCCACTTCTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601936 | ||||||||||||||||||||
Literature Links: |
PAWR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAWR - pro-apoptotic WT1 regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002583.2 | 925 | Silent Mutation | AAC,AAT | N,N 253 | NP_002574.2 | |
XM_006719435.2 | 925 | Silent Mutation | AAC,AAT | N,N 253 | XP_006719498.1 | |
XM_006719436.3 | 925 | Intron | XP_006719499.1 | |||
XM_017019377.1 | 925 | Silent Mutation | AAC,AAT | N,N 253 | XP_016874866.1 | |
XM_017019378.1 | 925 | Intron | XP_016874867.1 | |||
XM_017019379.1 | 925 | Intron | XP_016874868.1 |