Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCTATTCTTTCCCCGGAGCCA[A/G]TTCTCACAGGCCTGTGGATTAGCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613785 MIM: 120580 | ||||||||||||||||||||
Literature Links: |
C1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1R - complement C1r subcomponent | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001733.4 | 1999 | Silent Mutation | AAC,AAT | N,N 622 | NP_001724.3 | |
XM_017019915.1 | 1999 | Silent Mutation | AAC,AAT | N,N 636 | XP_016875404.1 |
C1S - complement component 1, s subcomponent | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |