Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGCAGGTGATGGCTTGATTCCT[C/T]GGGCTCTCATATAGGAGAGAAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CPNE8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPNE8 - copine 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153634.2 | 1655 | Missense Mutation | CAA,CGA | Q,R 542 | NP_705898.1 | |
XM_011537951.2 | 1655 | Intron | XP_011536253.1 | |||
XM_011537952.2 | 1655 | Intron | XP_011536254.1 | |||
XM_017018851.1 | 1655 | Missense Mutation | CAA,CGA | Q,R 542 | XP_016874340.1 | |
XM_017018852.1 | 1655 | Missense Mutation | CAA,CGA | Q,R 381 | XP_016874341.1 |