Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCACCTCGCGCTCAGCCGCCTT[A/C]TCGGGCCGCGGCGCCCAGGTGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 148059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT7 - keratin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005556.3 | 131 | Missense Mutation | TTA,TTC | L,F 17 | NP_005547.3 | |
XM_011538325.1 | 131 | Missense Mutation | TTA,TTC | L,F 17 | XP_011536627.1 | |
XM_017019294.1 | 131 | Intron | XP_016874783.1 |
LINC00592 - long intergenic non-protein coding RNA 592 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |