Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGCTACTTTCATCTTCTGAT[A/C]AGTAATTTAAATATTCCGAACAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616062 MIM: 614939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKLE2 - ankyrin repeat and LEM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015114.2 | 2850 | UTR 3 | NP_055929.1 | |||
XM_005266159.3 | 2850 | UTR 3 | XP_005266216.1 | |||
XM_005266160.1 | 2850 | UTR 3 | XP_005266217.1 | |||
XM_006719735.1 | 2850 | UTR 3 | XP_006719798.1 | |||
XM_011534787.2 | 2850 | Intron | XP_011533089.1 | |||
XM_011534788.1 | 2850 | Intron | XP_011533090.1 | |||
XM_017019076.1 | 2850 | UTR 3 | XP_016874565.1 |
PGAM5 - PGAM family member 5, mitochondrial serine/threonine protein phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |