Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAACAACAAGACATAAAGTACT[C/T]ATTATGGAATTTTGTCCATGTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604834 | ||||||||||||||||||||
Literature Links: |
TBK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBK1 - TANK binding kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013254.3 | 411 | Silent Mutation | CTC,CTT | L,L 84 | NP_037386.1 | |
XM_005268809.1 | 411 | Silent Mutation | CTC,CTT | L,L 84 | XP_005268866.1 | |
XM_005268810.1 | 411 | Silent Mutation | CTC,CTT | L,L 84 | XP_005268867.1 |