Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATAGCACAGCTGCCAGTAGACC[C/T]CCGGTCACCCTGAGGCTGGTGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCBP2 - poly(rC) binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098620.2 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_001092090.1 | |
NM_001128911.1 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_001122383.1 | |
NM_001128912.1 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_001122384.1 | |
NM_001128913.1 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_001122385.1 | |
NM_001128914.1 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_001122386.1 | |
NM_005016.5 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_005007.2 | |
NM_031989.4 | 638 | Silent Mutation | CCC,CCT | P,P 96 | NP_114366.1 |
PCBP2-OT1 - PCBP2 overlapping transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |