Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGAGTGAGTGTTGATGATACT[C/T]GGAAGATCTGGGGAAGCATGTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616082 | ||||||||||||||||||||
Literature Links: |
C12orf4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf4 - chromosome 12 open reading frame 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304811.1 | 1618 | Missense Mutation | CAA,CGA | Q,R 541 | NP_001291740.1 | |
NM_020374.3 | 1618 | Missense Mutation | CAA,CGA | Q,R 541 | NP_065107.1 | |
XM_005253716.1 | 1618 | Missense Mutation | CAA,CGA | Q,R 498 | XP_005253773.1 | |
XM_006718992.3 | 1618 | Intron | XP_006719055.1 |