Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGAGATGAAAAAAATGAGCAAC[A/G]ACACAGATTTAAAACTAGCCTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605862 | ||||||||||||||||||||
Literature Links: |
TMEM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM5 - transmembrane protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278237.1 | 410 | UTR 5 | NP_001265166.1 | |||
NM_014254.2 | 410 | Missense Mutation | CAA,CGA | Q,R 78 | NP_055069.1 | |
XM_005268562.3 | 410 | UTR 5 | XP_005268619.1 | |||
XM_005268563.3 | 410 | UTR 5 | XP_005268620.1 | |||
XM_017018686.1 | 410 | UTR 5 | XP_016874175.1 | |||
XM_017018687.1 | 410 | UTR 5 | XP_016874176.1 |