Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTGTAGCTAAAAGGATGTGTG[A/G]CGGAATCGTTTGGTAATGGCAGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600130 | ||||||||||||||||||||
Literature Links: |
APOBEC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBEC1 - apolipoprotein B mRNA editing enzyme catalytic subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304566.1 | 682 | Missense Mutation | CCA,TCA | P,S 220 | NP_001291495.1 | |
NM_001644.4 | 682 | Missense Mutation | CCA,TCA | P,S 220 | NP_001635.2 | |
NM_005889.3 | 682 | Missense Mutation | CCA,TCA | P,S 175 | NP_005880.2 |