Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCCTTTCTCTAGAGAGATGGAA[A/G]GAGAGGTATTGCTAGAAGGATCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR548C - microRNA 548c | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR548Z - microRNA 548z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF3 - Ras association domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178169.3 | Intron | NP_835463.1 | ||||
XM_006719345.3 | Intron | XP_006719408.1 | ||||
XM_011538195.2 | Intron | XP_011536497.1 | ||||
XM_017019182.1 | Intron | XP_016874671.1 |