Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCGCCCCACCGAGTTCGTCAA[C/G]CACTACTGCTGCGACAGCCACCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601284 | ||||||||||||||||||||
Literature Links: |
ACVRL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACVRL1 - activin A receptor like type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000020.2 | 404 | Missense Mutation | AAC,AAG | N,K 86 | NP_000011.2 | |
NM_001077401.1 | 404 | Missense Mutation | AAC,AAG | N,K 86 | NP_001070869.1 | |
XM_005269235.2 | 404 | Missense Mutation | AAC,AAG | N,K 86 | XP_005269292.1 | |
XM_011539008.1 | 404 | Missense Mutation | AAC,AAG | N,K 100 | XP_011537310.1 |