Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAGAAAAGCACCAACGTAAACA[A/G]TTACTTTTATTTTGAGTAAAAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600769 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN8 - tetraspanin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004616.2 | 1120 | UTR 3 | NP_004607.1 | |||
XM_006719583.3 | 1120 | UTR 3 | XP_006719646.1 | |||
XM_017019913.1 | 1120 | UTR 3 | XP_016875402.1 |