Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGTTAGTATAGGTTGGACGCTC[A/G]ATATCGAGGTTTCTACGACAGATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602529 | ||||||||||||||||||||
Literature Links: |
TUBA1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TUBA1A - tubulin alpha 1a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270399.1 | 1090 | Silent Mutation | ATC,ATT | I,I 219 | NP_001257328.1 | |
NM_001270400.1 | 1090 | Silent Mutation | ATC,ATT | I,I 184 | NP_001257329.1 | |
NM_006009.3 | 1090 | Silent Mutation | ATC,ATT | I,I 219 | NP_006000.2 |