Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTCTGTCGTGAGGCAGTGCGGAC[A/G]GGGACCCTCTGGGATTCTGCTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPB8 - heat shock protein family B (small) member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014365.2 | 287 | UTR 5 | NP_055180.1 |
LOC105370024 - uncharacterized LOC105370024 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |