Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCCCCACCGTCAATCCGTCCAC[C/T]GGAGAGGTCATCTGTCAGGTAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100650 | ||||||||||||||||||||
Literature Links: |
ALDH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH2 - aldehyde dehydrogenase 2 family (mitochondrial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000690.3 | 279 | Silent Mutation | ACC,ACT | T,T 61 | NP_000681.2 | |
NM_001204889.1 | 279 | Silent Mutation | ACC,ACT | T,T 61 | NP_001191818.1 |