Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605560 MIM: 605559 MIM: 609687 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXC-AS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXC-AS2 - HOXC cluster antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXC-AS3 - HOXC cluster antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXC10 - homeobox C10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017409.3 | 390 | Missense Mutation | GCC,TCC | A,S 98 | NP_059105.2 |
HOXC11 - homeobox C11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR196A2 - microRNA 196a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |